Avancé
SDS
Certificats
Données du fabricant
$0.00
Order Entry
Connectez-vous
/
Registre
Products
Services
Digital Solutions
Applications
Support
About
Saisie de commande
Statut de la commande
Votre compte
Panier
Canada
Saisie de commande
Statut de la commande
Votre compte
Panier
ContactUsLinkComponent
Domicile
Data Loggers
Accessories for iMini® Temperature Logger
Accessories for iMini® Temperature Logger
:
Cryopak Verification Technologies
Accessories for iMini® Temperature Logger
:
Cryopak Verification Technologies
Catalog No:
10033-810
10033-920
10033-952
Interface:
USB
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Disposable Petri Dishes
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International