Quick Press, PerkinElmer | Avantor
Quick Press, PerkinElmer
Quick Press with 7 mm Die Set
Catalog # CA97013-212
Supplier:   PerkinElmer
Fournisseur #:  01860436
Quick Press, PerkinElmer
Catalog # CA97013-212
Supplier:  PerkinElmer
Quick Press, PerkinElmer
Quick Press, PerkinElmer
Quick Press with 7 mm Die Set
Price
5 647,01 $
Log in pour la disponibilité
Each
Product Details & Documents
The quick press consists of a handpress, a 7 mm die set, and a metal collar.
  • Stainless steel construction
  • Ergonomic handle design
  • Easy to install
Micropellets can be prepared in the quick press by purchasing the optional 3 mm or 1 mm die. The KBr pellet is formed in a metal collar that fits into an adjustable pellet holder.
Spécifications
  • Désignation:
    Quick Press with 7 mm Die Set
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Adjustable Pellet Holder Adapter
    Adjustable Pellet Holder Adapter
    CA97013-508
    Log in pour la disponibilité
    Each - $243.88