ContactUsLinkComponent
Jacketed Pilot Plant Reactor Pressure Relief Manifold, Ace Glass Incorporated
Catalogue # 89097-262
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Jacketed Pilot Plant Reactor Pressure Relief Manifold, Ace Glass Incorporated
Catalogue # 89097-262
Fournisseur:  Ace Glass
Supplier Number:  10015-10
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
  • Description:
    Pressure relief manifold for ACE pilot plants
  • Cat. no.:
    89097-262
  • Supplier No.:
    10015-10
Spécifications
À propos de cet article
A pressure relief manifold for a jacketed, reaction vessel is designed to protect the vessel from breakage while isolated or disconnected from the circulator
Installed on the outlet side of the vessel,the manifold will reduce the liklihood of breakage caused by an excessive pressure differential exerted by the thermal transfer fluid. Complete system includes aluminum body, outlet fittings, hose, relief valve set at 10 psi, and pressure gauge. Fits 10 l and above.