ContactUsLinkComponent
EVE™ Automated Cell Counter, NanoEnTek
EVE™ Automated Cell Counter, NanoEnTek
Catalog # 10027-452
Supplier:  NANOENTEK USA INC.
UL Listed
undefined
EVE™ Automated Cell Counter, NanoEnTek
Catalog # 10027-452
Supplier:  NANOENTEK USA INC.
Supplier Number:  E1000
UL Listed
Specifications
  • Description:
    EVE™ Automated Cell Counter
  • Electrical:
    100–240V, 50/60Hz, 1.5A
  • Shipping weight:
    4.3 kg (9.5 lbs.)
  • W×D×H:
    27×20×19 cm
  • Shipping dimensions:
    38W × 30.5D × 38H cm
  • Weight:
    2.1 kg (4.6 lbs.)
  • Cat. no.:
    10027-452
  • Supplier No.:
    E1000
Specifications
About this item
The user-friendly EVE™ Automated Cell Counter features a benchtop design to enable simple, fast, automated cell counting and viability measurements within 20 seconds
EVE™ cell counting slides offer repeatable and reliable measurements and analysis. The unique design eliminates the use of coverslips and allows for exact volume control. This reduces the user’s risk of exposure to infectious or hazardous samples, and increases efficiency throughout laboratory procedures by eliminating the need for washing or autoclaving components. Each slide contains two enclosed chambers which can measure two different samples or perform replicates of the same sample.
The Cell Counter measures cell concentrations ranging from 1x104 to 1x107 cells/mL, and cells with sizes ranging from 5 to 60µm. The unit is also equipped with a clumping cell counting function to provide more accurate results (using EVE ™ PC software).
Certifications: CE marked. UL listed.
Ordering information: The EVE Automated Cell Counter includes a 110V power cord, a 12V adaptor, one box of Cell Coutning Slides (50 slides/100 counts), USB drive (2GB), User Manual, Quick Manual, and a one-year warranty. Additional Chamber Slides (10027-446) may be ordered seperately.
Cell Counting Slides
Product Information