Pocket Pro+, pH Tester, Hach
Pocket Pro+, pH Tester, Hach
Supplier:   Hach
Pocket Pro+, pH Tester, Hach
Supplier:  Hach
Certificates
Pocket Pro+, pH Tester, Hach
1 Options Available Below
Product Details & Documents
Hach Pocket Pro and Pro+ are engineered to deliver accurate results. Calibration functions for ongoing testing, diagnostics with select models and a stabilized sample lock ensure you get the right reading. You also get replaceable batteries for convenient field use, and a large, easy-to-read LCD screen.
Confidence in results; to maximize accuracy all testers are factory calibrated, select models offer user-calibration, and pH models have performance diagnostics.
Auto-Buffer Recognition. ATC. Selectable Temp (°C or °F). Auto-Off. Reading Hold. Waterproof (IP 67 and floats). Quick-Release Lanyard (included). Transparent Sensor Cap.
Save time and money; easy-to-replace AAA batteries and convenient sample cup save time on maintenance and measurements. Ease of use; intuitive user interface guides users through the calibration and measurement process with a large, easy-to-read LCD screen.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 89508-374
  • CA89508-374
  • pH range:
  • 0.0 - 14.0
  • 0.0 - 14.0
  • pH resolution:
  • 0.1
  • 0.1
  • pH accuracy:
  • ±0.02 pH
  • ±0.02 pH
  • pH calibration:
  • 3 points; customer-defined 2 point
  • 3 points; customer-defined 2 point
  • Sensor type:
  • Single-Junction (electrode/ sensor)
  • Single-Junction (electrode/ sensor)
  • Battery type:
  • AAA×4
  • AAA×4
  • Battery life:
  • >280 hrs
  • >280 hrs
  • Operating temperature range:
  • 0…50 °C
  • 0…50 °C
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Pocket Pro+, pH
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    89508-374
    pH range
    0.0 - 14.0
    pH resolution
    0.1
    pH accuracy
    ±0.02 pH
    pH calibration
    3 points; customer-defined 2 point
    Sensor type
    Single-Junction (electrode/ sensor)
    Battery type
    AAA×4
    Battery life
    >280 hrs
    Operating temperature range
    0…50 °C
  • Pocket Pro+, pH
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $997.64
    $997.64
    Each
       $997.64
    Supplier Number: 9532000
    Spécifications:
    More
    Specifications
    Cat. No.
    CA89508-374
    pH range
    0.0 - 14.0
    pH resolution
    0.1
    pH accuracy
    ±0.02 pH
    pH calibration
    3 points; customer-defined 2 point
    Sensor type
    Single-Junction (electrode/ sensor)
    Battery type
    AAA×4
    Battery life
    >280 hrs
    Operating temperature range
    0…50 °C
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Replacement sensor, Pocket Pro+, pH
    Replacement sensor, Pocket Pro+, pH
    CA89508-376
    Log in pour la disponibilité
    Each - $423.70