SP Bel-Art Burette Clamp, Spring Action, Bel-Art Products, a part of SP
Catalog # 47750-930
Supplier: Bel-Art Products, a Part of SP
Specifications
- Description:Burette Clamp, Spring Action
- Cat. no.:47750-930
- Overall Length:22 cm (9")
- Supplier No.:183540000
Specifications
About this item
Firmly holds test-tubes, burettes and other labware in place.
- Spring-action design
- Stainless steel construction
- Tightened with a flat wing-head screw
- Covered in rubber tubing for increased friction
The spring action, stainless steel jaws have a flat wing-head screw for tightening, are covered with rubber tubing for increased friction and a soft grip, and have an opening range of 0 to 25 mm (1").