Biopsy Cassettes, FastLoad Taped Stacks, Closed Lid, Azer Scientific
FastLoad Biopsy Taped Cassettes feature a recessed hinged lid that can be opened and closed as often as necessary and always relocks securely.
- Attached lid
- Cassettes taped for automated systems
- Histology-solvents resistant
- Three frosted writing areas
The lid is attached and in the closed position.
Cassettes are molded from a high-density polymer.
: Cassettes are packaged in taped stacks of 40. Each case contains 50 stacks for a total of 2000 per case.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together