À propos de cet article
FastLoad Tissue Taped Cassettes feature a recessed hinged lid that can be open and closed as often as necessary and always relocks securely.
- Attached and closed lid
- Recessed, hinged lid that securely refastens
- Three frosted writing areas
- High-density polymer resistant to chemical actions
- Cassettes taped for automated systems
- Anterior writing area at 45° angle
Cassettes are molded from a high-density polymer that is resistant to solvents
The recessed lid allows for more cassettes to be stacked in auotmated systems and in storage. It is attached and in the closed position.
They are taped for use in automated systems.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.