Embedding Cassettes, FastLoad Mesh Taped Stacks, Closed Lid, Azer Scientific
Supplier: VWR International
Certificates
About this item
FastLoad Mesh Taped Cassettes feature 0.38 mm square openings.
- Back Tab for easy handling of cassettes
- Square openings at 0.38 mm
- Three frosted writing areas
- Cassettes taped for automated systems
- Recessed, hinged lid
- Anterior surface at a 45° angle
These cassettes have large anterior slots to allow for increased fluid exchange.
Each cassette also features a recessed hinged lid.
Packaging: Cassettes are packaged in taped stacks of 40. Each case contains 50 stacks for a total of 2000 per case.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together