SP Bel-Art Precision Plastic Dial Calipers, Bel-Art Products, a part of SP
Catalog # 13196-070
Supplier: Bel-Art Products, a Part of SP
Specifications
- Description:Dial Calipers
- Overall length:22.2 cm (8¾")
- Measuring system:Metric Only
- Cat. no.:13196-070
- Supplier No.:134160001
Specifications
About this item
Dial type calipers measure up to 150 mm in 0.1 mm increments.
- Dial-type design
- Non-conducting
- Corrosion resistant
- Lightweight for easy handling
- Rigid plastic construction
A 33 mm (1.3") diameter dial is rotatable beneath the pointer allowing for ‘differential’ measurements. Black numbers on a white background afford effortless readability. These calipers are non-conducting, corrosion resistant, and lightweight for easy handling. Molded-in rigid plastic, they will not scratch like metal calipers can.