Ecosense DO200A Handheld Dissolved Oxygen Meters, YSI
Fournisseur: YSI
Certificats
À propos de cet article
The DO200A is part of the EcoSense® family of products. The DO200A dissolved oxygen instrument offers fast dissolved oxygen response times in wastewater, surface water, and aquaculture applications.
- 1-year instrument, DO electrode and cable warranty
- IP 67 waterproof housing
- Screw-on cap membrane
- Low battery indicator with 500 hour battery life
- Automatic shutoff after 30 minutes of inactivity
- Salinity, pressure and temperature compensation
- 4- or 10- meter cable lengths available, BOD probe available
- 50 data set reviewable memory
Calibrates easily in a saturated air environment and displays in % or ppm (mg/L) values, giving you the best DO data quality possible. Available with 4- or 10-meter cable lengths. Audio feedback, all keys. Calibration back up. Grey color.
The DO200A (gray case) directly replaces the DO200 (blue case).
Pressure compensation: 600 - 1100 millibar (450 - 825 mmHg)
Certifications: CE compliance. IP67 waterproof rating.
Ordering information: Instrument and cables sold seperately unless ordering as a kit. Kits include (89423-010, 89423-012, or 89423-014): instrument, cable (1-, 4-, or 10-meter), probe and carrying case. One 9V battery included. 1-year instrument and electrode warranty.
Spécifications
- Catalog No:
- 89423-006
- 89423-012
- 89423-014
- 89423-010
- DO range:
- 0 - 20 ppm (mg/L)
- 0 to 20 ppm (mg/L)
- 0 to 20 ppm (mg/L)
- 0 to 20 ppm (mg/L)
- DO resolution:
- 0.1% air saturation
- 0.1% air saturation
- 0.1% air saturation
- 0.1% air saturation
- DO accuracy:
- ±2% of the reading or ±0.2 ppm, whichever is greater
±2% of the reading or
±2% air saturation, whichever is greater - ±2% of the reading or ±0.2 ppm, whichever is greater
±2% of the reading or
±2% air saturation, whichever is greater - ±2% of the reading or ±0.2 ppm, whichever is greater
±2% of the reading or
±2% air saturation, whichever is greater - ±2% of the reading or ±0.2 ppm, whichever is greater
±2% of the reading or
±2% air saturation, whichever is greater
- DO saturation:
- 0 - 200%
- 0 to 200%
- 0 to 200%
- 0 to 200%
- DO salinity compensation:
- 0.0 - 40.0 ppt
- 0.0 - 40.0 ppt
- 0.0 - 40.0 ppt
- 0.0 - 40.0 ppt
- Temperature resolution:
- 0.1 ºC
- 0.1 ºC
- 0.1 ºC
- 0.1 ºC
- Temperature ATC probe:
- Thermistor, 10kΩ /25°C
- Thermistor, 10kΩ /25°C
- Thermistor, 10kΩ /25°C
- Thermistor, 10kΩ /25°C
- Batteries:
- One 9V, 500 h
- One 9V, 500 hours
- One 9V, 500 hours
- One 9V, 500 hours
- Memory:
- 50 Data Set Reviewable Memory
- 50 Data Set Reviewable Memory
- 50 Data Set Reviewable Memory
- 50 Data Set Reviewable Memory
- Dimensions:
- 78W x 184L mm (3.1 x 7.25")
- 78W x 184L mm (3.1 x 7.25")
- 78W x 184L mm (3.1 x 7.25")
- 78W x 184L mm (3.1 x 7.25")
- Weight:
- 272 g (0.6 lb.) with battery
- 272 g (0.6 lb.) with battery
- 272 g (0.6 lb.) with battery
- 272 g (0.6 lb.) with battery
- Operating temperature range:
- 0...50 °C
- 0...50 °C
- 0...50 °C
- 0...50 °C
- Operating humidity range:
- up to 95%
- up to 95%
- up to 95%
- up to 95%