SP Bel-Art Safe-Lab™ Joint Stirrer Bearing for Tapered Joints, Bel-Art Products, a part of SP
70680-452
- Cat. no.:70680-452
- ST Joint Size:24/40
- Supplier No.:203201660
The body is Teflon® PTFE with a Viton® O-Ring and is designed for use with 10 mm stirring rods.
- Threaded polypropylene locking compression cap
- Easy removal with extracting nut above the ground joint
- Not autoclavable