ContactUsLinkComponent
SpecCheck Secondary Gel Standards Set, DPD Chlorine - MR, Hach
Catalog # CA11014-246
Supplier:  Hach
CAS Number:  
undefined
SpecCheck Secondary Gel Standards Set, DPD Chlorine - MR, Hach
Catalog # CA11014-246
Supplier:  Hach
Supplier Number:  2980500
CAS Number:  
Specifications
  • Description:
    SpecCheck Secondary Gel Standards Set, DPD Chlorine - MR
  • Cat. no.:
    CA11014-246
  • Supplier No.:
    2980500
Specifications
About this item
Chlorine SpecCheck Secondary Standards Kit for Mid Range DPD Chlorine Methods. Contains a blank vial and 3 colored vials at ~0.2, ~1.5, and ~3.5 mg/L Cl2. SpecCheck Standards are computer-formulated and have a two-year shelf life from date of manufacture. For use only with Mid Range DPD chlorine methods. Not for use with Low Range methods (including LR/HR Pocket Colorimeter II).
  • Solid gel standards
  • No preparation required
  • Ideal for insturment verification in the lab and in the field
  • NIST traceable
Ready-to-use SpecCheck Secondary Standards Kits include four 10 ml (1") sealed glass cells with a blank and three different standards within the typical testing range, instructions, plastic case and Certificate of Analysis.
Ordering information: Product has guaranteed performance for two years. This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.