EvoControl 400 Cleanroom Wipes, Vileda Professional-FHP
Fournisseur: Vileda Professional - FHP
Certificats
À propos de cet article
Maximize contamination control with Vileda® wipes! The microfiber EvoControl cleanroom wipes fulfill the highest demands for cleanroom wiping, plus EvoControl provides revolutionary wiping results thanks to its unique microfiber structure.
- EvoControl 400 is a disposable liquid cleaning wipe designed for contamination removal
- It is pre-laundered, laser cut and designed for the removal of acids and chemical spills
Evolon microfiber cleans glass and stainless steel without leaving residues or streaking and also has a low particle release while in use. The EvoControl cleanroom microfiber is 10 times finer than standard knitted PES cleanroom wipes, removing contamination more thoroughly along with maximizing particle pick up and bacteria removal. The cloths laser cut sealed edge are melted together for a perfect clean edge, which prevents particle release.
Frequently Bought Together