Ohaus® Starter Electrodes
Ohaus® Starter Electrodes
Supplier:   Ohaus
Ohaus® Starter Electrodes
Supplier:  Ohaus
Certificates
Ohaus® Starter Electrodes
Ohaus® Starter Electrodes
7 Options Available Below
Product Details & Documents
The Ohaus® Starter Series of electrodes include pH, reference, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO) and temperature electrodes.
  • Can be used in conjunction with Ohaus® Starter Series
The Ohaus® Starter Series of electrodes include pH, reference, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO) and temperature electrodes that can be used in conjunction with Ohaus® Starter Series 2100 and 3100 bench and Ohaus® Starter 300 Series Portable meters.
Ordering information: Ohaus® 2100 & 3100 Starter Bench Meter Compatible Electrodes are ST210, ST320, STTEMP30. Ohaus® 3100C Starter Bench Meter Compatible Electrodes are STCON3. Ohaus® 300 Starter Portable Meter Compatible Electrodes are ST210, ST320, STTEMP30. Ohaus® 300C Starter Portable Meter Compatible Electrodes are STCON3. Ohaus® 300D Starter Portable Meter Compatible Electrodes are STTEMP30, STD011.
Product Family Options
Informations sur le produit
  • Désignation
    For Use With
    Availability
    Price
  • ST210: General purpose 2-in-1 plastic gel refillable pH electrode
    Series 300, 2100, 3100
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $135.50
    $135.50
    Each
       $135.50
    Supplier Number: 83033966
  • ST320: General purpose 3-in-1 plastic gel pH electrode
    Series 300, 2100, 3100
    Discontinued
    Supplier Number: 83033967
  • ST310: General purpose 3-in-1 plastic refillable pH electrode
    Series 300, 2100, 3100
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $325.10
    $325.10
    Each
       $325.10
    Supplier Number: 83033965
  • ST230: General purpose glass-body refillable pH electrode
    Series 300, 2100, 3100
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $282.54
    $282.54
    Each
       $282.54
    Supplier Number: 83033968
  • ST210: General purpose 2-in-1 plastic gel refillable pH electrode
    Series 300, 2100, 3100
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $151.48
    $151.48
    Each
       $151.48
    Supplier Number: 83033966
  • ST320: General purpose 3-in-1 plastic gel pH electrode
    Series 300, 2100, 3100
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $325.10
    $325.10
    Each
       $325.10
    Supplier Number: 83033967
  • STCON3: 4-Ring conductivity electrode
    Series 300C, 3100C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $614.20
    $614.20
    Each
       $614.20
    Supplier Number: 83033972
  • STD011: Galvanic DO electrode
    Series 300D
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $308.00
    $308.00
    Each
       $308.00
    Supplier Number: 30031639
  • STTEMP30: Temperature electrode
    Series 300, 300D, 2100, 3100
    Discontinued
    Supplier Number: 83033970
Accessories
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • Maintenance kit
    Maintenance kit
    77679-490
    STDO11: DO electrode
    Log in pour la disponibilité
    Each - 186,24 $