Micro Reaction Vial Caps, Ace Glass Incorporated
Threaded cap with or without drilled hole (solid) which includes PTFE faced silicone rubber septum and one o-ring for making “O-Ring-CAP-SEAL.” For use with Microscale Reaction Vials.
Frequently Bought Together