CDC866T Conductivity Cell, Hach
Catalogue # CA89501-078
Fournisseur: Hach
Spécifications
- Description:General Purpose Conductivity Cell, 4-Pole, Platinised, Sensor
- Cat. no.:CA89501-078
- Supplier No.:E61M015
Spécifications
À propos de cet article
CDC866T Conductivity Cell
4-pole. Platinised. Temperature Sensor. Removable Tube. MAB6 plug (Radiometer Analytical).