Digital Reactor Blocks, 21×16 mm Vial Wells/4×20 mm Vial Wells, DRB200
Fournisseur: Hach
Certificats
À propos de cet article
The simple solution for standard and special digestions.
- Easy-to-use and fast
- Safe to operate
- Versatile
- Accommodates most test vials
- Select the dual block model for simultaneous digestions
Applications: Boiler water, industrial wastewater, municipal water, pre-treatment, steam cycle.
Certifications: CE, GS, cTUVus.
Delivery: Includes: Digital block reactor, power cord and operating manual.
Spécifications
- Catalog No:
- CA11014-310
- Heat range:
- 20...150 °C in 10 min.
- Operating temperature range:
- 10...45 °C
- Temperature range:
- 37...165 °C (99...329 °F)
- Temperature stability:
- ±2 °C
- Weight:
- 2.8 kg (6.17 lbs.)