Digital Reactor Blocks, 30×16 mm Vial Wells, DRB200
Fournisseur: Hach
Certificats
À propos de cet article
The simple solution for standard and special digestions.
- Easy-to-use and fast
- Safe to operate
- Versatile
- Accommodates most test vials
- Select the dual block model for simultaneous digestions
Applications: Boiler water, industrial wastewater, municipal water, pre-treatment, steam cycle.
Delivery: Includes: Digital block reactor, power cord and operating manual.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- CA11014-308
- Temperature range:
- 37...165 °C (99...329 °F)
- Temperature stability:
- ±2 °C
- Weight:
- 2.0 kg (4.40 lbs)