TruCLEAN II Compact Flat Mopping System, Perfex
Fournisseur: Perfex
Certificats
À propos de cet article
Designed for small area cleaning and disinfecting
Compatible with gamma, ETO and autoclave sterilization – up to 250°F (121°C) for 30 minutes.
Includes 36-liter bucket with 3”casters, waste containment bucket, stainless steel wringer, stainless steel mop frame and polymer mop handle. Captures and isolates soiled contaminants from adulterating cleaning and disinfecting agents. Embossed graduations in English and Metric for accurate measure. Clean floors, walls and ceilings with this efficient, compact mopping system.
Frequently Bought Together