Corning® LSE™ Low Speed Orbital Shaker, Corning
Catalog # 89133-524
Supplier: Corning
Specifications
- Max. capacity:11.8 x 11.8" Platform
- Electrical:120V
- Dimensions:10 x 12.5 x 6.5"
- Weight:12 lbs.
- Cat. no.:89133-524
- Speed:3 - 60 rpm
- Supplier No.:6780-FP
Specifications
About this item
The Corning LSE Low Speed Orbital Shaker includes a 30x30cm platform and is ideal for staining and destaining gels, washing blots and general mixing applications
Speed can be set from 3 to 60 rpm and the timer can be set for up to 2 hours or for continuous operation. Analog control panel.
Certifications: CE Compliant, CSA Compliant.