Corning® LSE™ Orbital Shaker, Corning
89133-514
: Corning
Product available on GSA Advantage®
- Max. capacity:4 x 1 L
- Description:Orbital Shaker
- Electrical:120V
- Dimensions:10 x 12.5 x 5"
- Weight:15 lbs.
- Cat. no.:89133-514
- Speed:20 - 300 rpm
- Supplier No.:6780-NP
The Corning LSE Orbital Shaker meets a variety of shaking and mixing applications
A powerful brushless motor and counter-balanced shaker mechanism provide smooth operation for years of trouble-free use. Different platforms (order separately) are available and are easily interchangeable. Digital control panel.
: CE Compliant, CSA Compliant.