TLG® Cylinders, To Contain, Class A, White Scale, with Reverse Graduations and Bumper, with LOT Calibration Certificate, SATI Glassware
Fournisseur: CHEM SCIENCE INC CA
Certificats
À propos de cet article
TLG® Brand Cylinders come with reverse metric scales
The "TC" (to contain) appears on each cylinder and indicates the cylinder is calibrated to contain. Hexagonal bases are flat ground for added stability. Safety bumpers are supplied with sizes 25mL through 2000 mL. Designed from ASTM Specification E1272, Class A Certified.
Certifications: ASTM Specification E1272, Class A certified.
Spécifications
- Catalog No:
- CA10823-762
- CA10823-760
- CA10823-554
- CA10823-552
- CA10823-550
- Form:
- Tall
- Tall
- Tall
- Tall
- Tall