ContactUsLinkComponent
TLG® Griffin Low Form Beakers with Double-Capacity Scale, SATI Glassware
  CA10823-748
 : 229.202.11
undefined
TLG® Griffin Low Form Beakers with Double-Capacity Scale, SATI Glassware
  CA10823-748
 :  229.202.02A
 : 229.202.11
Select an Option
108 mm
Select an Option
50 - 1000 ml
Select an Option
1000 ml (34 oz.)
Select an Option
156 mm
Select an Option
CA10823-748
Select an Option
229.202.11
TLG® Brand Beakers meet ASTM E-960 Specifications for classification, design, capacity & dimensions, and markings
  • Easy-pour spouts
  • White, permanent graduations
  • Large marking squares
  • Made from high-quality borosilicate glass with a low coefficient of expansion
Manufactured with uniform wall thickness resulting in the optimum balance between thermal shock resistance and mechanical strength. 20mL through 4000mL beakers have double-graduated metric scales. Accuracy: ±5% of full capacity.
 : ASTM E-960.
Frequently Bought Together