BugBuster® Protein Extraction Reagent, MilliporeSigma
BugBuster® Protein Extraction Reagent is formulated for the gentle disruption of the cell wall of E.coli, resulting in the liberation of soluble protein.
It provides a simple, rapid, low cost alternative to mechanical methods such as French Press or sonication for releasing expressed target proteins in preparation for functional studies or purification.
Frequently Bought Together