Variable Volume Pipettes, Hach
Supplier:   Hach
Variable Volume Pipettes, Hach
Supplier:  Hach
Certificates
Variable Volume Pipettes, Hach
Variable Volume Pipettes, Hach
2 Options Available Below
Product Details & Documents
Hach's variable volume single-channel, air-displacement pipettes feature robust design, comfortable ergonomics, slim-line tip ejection, and single-handed operation.
Outstanding accuracy with adjustments in 50 L increments (0.05 ml).
 : Supplied with calibration report and maintenance tool.
Product Family Options
Informations sur le produit
  • Désignation
    Volume
    Availability
    Price
  • Variable Volume Pipet
    0.2 - 1.0 ml
    Discontinued
    Supplier Number: -
  • Variable Volume Pipet
    1.0 - 5.0 ml
    Discontinued
    Supplier Number: -
  • Variable Volume Pipette Set with Tips
    1× each 0.2 - 1.0 ml and 1.0 - 5.0 ml
    Discontinued
    Supplier Number: -
  • Variable Volume Pipet
    0.2 - 1.0 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,346.66
    $1,346.66
    Each
       $1,346.66
    Supplier Number: BBP078
  • Pipette, Variable Volume
    1.0 - 5.0 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,358.17
    $1,358.17
    Each
       $1,358.17
    Supplier Number: BBP065
  • Variable Volume Pipette Set with Tips
    1× each 0.2 - 1.0 ml and 1.0 - 5.0 ml
    Discontinued
    Supplier Number: -