Anti-MAP3K7 Rabbit Polyclonal Antibody
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
TAK1 Antibody: TAK1 (TGF-beta activated protein kinase 1) is a widely expressed enzyme originally identified as a mitogen-activated protein kinase kinase kinase (MAP3K7). It has since been shown to mediate various intracellular signaling pathways, such as those induced by TGF-beta and members of the Toll-IL-1R (TIR) superfamily, thus acting as an intermediate in both proliferative and innate and adaptive immune responses. TAK1 is normally present in cells in a complex with TAK1-binding protein 1 (TAB1) and either TAB2 or TAB3. Association with TAB1 triggers TAK1 kinase activity by inducing TAK1 autophosphorylation, while TAB2 and possibly TAB3 may contribute to SAPK2a/p38a-mediated feedback inhibition of TAK1 activity.
Type: Primary
Antigen: MAP3K7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Spécifications
- Taille:0.1 mg
- Western blot:Yes
- Clonality:Polyclonal
- Form:Liquid
- ImmunoFluorescence:Yes
- Storage temperature:TAK1 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Antigen name:mitogen-activated protein kinase kinase kinase 7
- Gene ID:O43318
- Immunogen:TAK1 antibody was raised against a synthetic peptide corresponding to 14 amino acids near the amino-terminus of human TAK1 isoform a. The immunogen is located within amino acids 40 - 90 of TAK1.
- Tested applications:E, WB, IF
- Storage buffer:TAK1 Antibody is supplied in PBS containing 0.02% sodium azide.
- Purification:TAK1 Antibody is affinity chromatography purified via peptide column.
- Antibody type:Primary
- Isotype:IgG
- Host:Rabbit
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Cat. No.:89415-726
- Reactivity:Human|Rat|Mouse
- Antigen synonyms:MAP3K7
- Conjugation:Unconjugated
- Antigen symbol:MAP3K7
- ELISA:Yes
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



