Sampling Swab for Environmental Monitoring, Sterile Pack, BD Diagnostics
Catalog # CA-90001-120
Supplier: BD
Specifications
- Description:Sterile Pack Swab
- Cat. no.:CA-90001-120
- Supplier No.:220518
Specifications
About this item
Each sterile pack sampling swab unit is ready-to-use and comprised of a peel open pouch containing an environmental sampling swab in a tube with approximately 10 mL rinse solution
- Ready-to-use sterile swab in a pre-filled tube of rinse solution
- Use for surface and equipment sampling
- Double-wrapped and gamma irradiated
The pre-filled rinse solution is a general-purpose isotonic solution with neutralizing agents for the maintenance of microorganisms and the neutralization of disinfectants. The double pouch design is gamma-irradiated.
Designed to sample a variety of surface textures and equipment with the advantage of sampling hard-to-reach areas such as equipment crevices and inside pipe work.