ContactUsLinkComponent
Tris-Buffered Saline with Tween Solution (10X), Genetex
Tris-Buffered Saline with Tween Solution (10X), Genetex
Catalog # 89306-978
Supplier:  Genetex
CAS Number:  
Tris-Buffered Saline with Tween Solution (10X), Genetex
Catalog # 89306-978
Supplier:  Genetex
Supplier Number:  GTX30976
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Tris-Buffered Saline (10X)
  • Cat. no.:
    89306-978
  • Supplier No.:
    GTX30976
Specifications
About this item
Dilute this 10X buffer 10 times, mix well, and store 1 X buffer solution at room temperature or 2-8oC
Liquid form. Store at room temperature. If stored at 2°C-8°C, some sodium chloride will be precipitated, if this occurs, bring the buffer to room temperature and mix well till all sodium chloride is dissolved.