ContactUsLinkComponent
PBS Phosphate Buffered Saline Tablets, Genetex
  89284-786
 :  Genetex
 :  
PBS Phosphate Buffered Saline Tablets, Genetex
  89284-786
 :  Genetex
 :  GTX48887
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    PBS Tablets Sufficient for 100 mL
  • Size:
    100 Tablets
  • Cat. no.:
    89284-786
  • Supplier No.:
    GTX48887
Pre-measured Phosphate Buffered Saline Tablets
Each tablet prepares a 1X PBS solution when dissolved in 100 mL of ddH2O. For in vitro research use only. Not intended for any diagnostic or therapeutic purpose. Not suitable for human or animal consumption. Store at room temperature.