SP Bel-Art Poxygrid® Microliter Pipettor Racks for Gilson Pipetman®, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
À propos de cet article
Choose from three styles of epoxy-coated aluminum rack to hold Gilson Pipetman® pipettors.
- Epoxy-coated aluminum rack construction
- Extended base with raised lip
- Suitable for wall mounting or free standing
Racks provide storage and easy access for up to 10 pipettors, plus up to two Pipet-Aid® pipettes. Rack 53577-148 is designed for desktop storage, while rack 161256-684 is designed for wall mounting with two holes for screw attachment to the wall and double faced tape (included). Rack 13197-158 has an extended base with a raised lip to hold plastic tip boxes, and is suitable for wall mounting or for free standing use on a bench.
Frequently Bought Together