SP Bel-Art Stirring Paddle, Bel-Art Products, a part of SP
13197-418
- Description:Stirring Paddle
- Cat. no.:13197-418
- Supplier No.:377700000
Designed for mixing liquids in large vats and tanks, this high-density polyethylene stirring paddle has a 91 cm (3') handle.
- HDPE construction
- Designed for use with large vats and tanks
- Long handle extends reach and provides extra safety
The paddle measures 7.6x15.2 cm (3x6"), and the handle has a 25 mm (1") diameter.