SP Bel-Art Pipette Tray Rack, Bel-Art Products, a part of SP
Catalogue # 53574-100
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Description:Pipette Tray Rack
- Dimensions:286L x 216W x 29H mm (11¼ x 8½ x 1⅛")
- Cat. no.:53574-100
- Supplier No.:189400000
Spécifications
À propos de cet article
This styrene tray is useful in lab drawers or on a benchtop for soaking, draining, or storing pipettes.
- Styrene construction
- Scalloped rims hold different pipette sizes
- Compact design
Scalloped rims hold 16 small or 7 large pipettes. Center support holds 12 short pipettes.