Solid Cable Ties, Antylia Scientific | Avantor
Solid Cable Ties, Antylia Scientific
Solid Cable Ties, Antylia Scientific
Supplier:  Antyila Scientific
Certificates
Solid Cable Ties, Antylia Scientific
Solid Cable Ties, Antylia Scientific
2 Options Available Below
Product Details & Documents
Solid Cable Ties won’t corrode or stretch. They are made from injection-molded plastic.
  • Simply bend tip and pull through locking mechanism to secure
  • Ties won't lose tension, even in vibrating, oily, or wet environments
  • Ties withstand temperatures to 85 °C (185 °F), with a melting point of 257 °C (495 °F)
Ties are injection-molded of 6/6 nylon that won’t corrode or stretch. Durable locking mechanism holds securely even when used to bundle vibrating or flexing materials. Ties continue to hold tension even in oily spray or wet environments. They can be easily applied by hand or use a tensioning tool (tools must be ordered separately).
Ties are UL-component recognized, and are self-extinguishing as per UL 94 Flam. Class: V-2 (models 06830-85 and -95 have a UL flame rating of UL94-HB). All solid design cable ties ('A', and 'E' meet military specification MS3367 except the extra-heavy duty models (06830-42 and -99). Ties withstand temperatures to 185° F (85° C) and have a melting point of 257 °C (495 °F); except for barbed design cable ties which withstand temperatures to 75 °C (167 °F)and have a melting point of 218 °C (425 °F). All ties remain flexible to –40 °C. Efficient range of ties may be higher or lower in specific applications.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Availability
    Price
  • Nylon cable tie wraps
    13.9 cm (5.5")
    Discontinued
    Supplier Number: -
  • Nylon cable tie wraps
    13.9 cm (5.5")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1,000
    Pack of 1,000 - $144.41
    $144.41
    Prix promotionnel pour les nouveaux clients
    Pack of 1,000
       $144.41
    Supplier Number: 06830-54
  • Nylon cable tie wraps
    19 cm (7.5")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1,000
    Pack of 1,000 - $227.33
    $227.33
    Prix promotionnel pour les nouveaux clients
    Pack of 1,000
       $227.33
    Supplier Number: 06830-56