Single Metric Scale Cylinders with Bumper, Kimble Chase
89000-968
- Description:Starter Pack of 5 Single Metric Scale Cylinders with Bumpers, Class B
- Form:Short
- Cat. no.:89000-968
- Supplier No.:20024-01
An assortment of five popular sized graduated cylinders (one each) in sizes of 10, 25, 50, 100 and 250 mL
The 10 mL cylinder has graduations from 1 to 10 mL; the 25 mL cylinder has graduations from 3 to 25 mL; the 50 mL cylinder has graduations from 3 to 50 mL; the 100 mL cylinder has graduations from 5 to 100 mL; the 250 mL cylinder has graduations from 10 to 250 mL.
Ideal for start-up requirements and customers who need a variety of cylinders, but have limited lab space or glassware needs.