Color Frosted Ground Edge Microscope Slide, Globe Scientific
Supplier: Globe Scientific
Certificates
About this item
Color frosted microscope slides are offered in 10 bright, attractive colors.
- Dimensions: 25×75×1 mm
- Thickness: 1.1 mm
- Color frosted one end, one side
- Packaging: 72/box, 20 boxes/case (10 gross)
Slides are pre-cleaned and supplied ready for use.
All slides are printed with 'Globe' under the frosting area for orientation purposes. These slides feature safety corners, which provide slightly clipped corners for comfortable handling and to prevent the risk of finger and hand cuts.
Frequently Bought Together