UVP UV/White Light Transilluminators, Analytik Jena
UV/White Transilluminators house white light and UV working areas side by side for multiple applications
- Increased ultraviolet output and intensity
- No light flicker
- Wider intensity range with variable selection (select models)
- Fast lamp start up
- Universal power supply
- Reduced electrical consumption
- Longer tube life
- Lighter weight
- Improved gel visualization and gel documentation
Caution: Transilluminators are powerful sources of ultraviolet radiation which can cause serious damage to unprotected eyes and skin. Read the ultraviolet Caution Information before operating any transilluminators.
New electronic ballast and cutting-edge construction. Single intensity style.
Frequently Bought Together