Slide Mailer, Screw-Cap, Globe Scientific
Catalog # 89510-784
Supplier: Globe Scientific
Specifications
- Color:Red Screw Cap
- Description:Slide Mailer for 4 Slides
- Cat. no.:89510-784
- Supplier No.:513068
Specifications
About this item
This screw-cap slide mailer is produced from Polypropylene (PP) and can hold 4 standard sized microscope slides (25 x 755 mm or 3 x 1")
The inner slots keep slides separated to avoid cross-contamination during shipping or storage.