Spherical Jacketed Reaction Flask, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Rugged spherical jacketed reaction flask with Duran style flat flange with O-ring groove, for use with Ace Glass quick release clamp
Each flask is supplied with one CAPFE (PTFE encapsulated silicone) o-ring. Plain silicone O-rings are also available. The 12 liter size is supplied with capacity reference graduations. Inlet and outlet are size 28/15 O-ring ball joints, sealed tangentially.
Frequently Bought Together