Xylene Substitute, Electron Microscopy Sciences | Avantor
Xylene Substitute, Electron Microscopy Sciences
Xylene Substitute, Electron Microscopy Sciences
Certificates
Xylene Substitute, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This EMS Xylene Substitute solution is specially designed for use in processing and staining histological/cytological specimens. It is essentially odorless with low toxicity, less irritating to the skin than xylene- and d-limonene-based clearants. It can be substituted for xylene and can be used with almost all tissue processors and automated slide stainers.
  • Minimal tissue shrinkage
  • Dries slowly with no residue
  • Less toxic than xylene
  • Soluble with alcohol, paraffin embedding media, and mounting media
  • Biodegradable
Xylene substitute for histology and cytology is a safer alternative to xylene. Can be used as a solvent and clearing agent, as well as for dissolving paraffin waxes, glues and adhesives. It can be used in all procedures that require xylene.
Product Family Options
Informations sur le produit
  • Désignation
    Type de conditionnement
    Taille
    Availability
    Price
  • Xylene substitute
    Bottle
    1 gal.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (1US Gal)
    Each (1US Gal) - $337.64
    $337.64
    Each (1US Gal)
       $337.64
    Supplier Number: 23410-01
  • Xylene substitute
    Bottle
    4×1 gal.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $1,265.92
    $1,265.92
    Log in pour la disponibilité
    Each
    Each - $1,265.92
    $1,265.92
    Case of 4
       $1,265.92
    Each
       $1,265.92
    Supplier Number: 23410-04
  • Xylene substitute
    Bottle
    5 gal.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (5US Gal)
    Each (5US Gal) - $1,264.94
    $1,264.94
    Each (5US Gal)
       $1,264.94
    Supplier Number: 23410-05