DURAEDGE® Standard Microtome Blades, Crescent®
Fournisseur: CRESCENT MANUFACTURING
Certificats
À propos de cet article
Standard disposable microtome blades for sectioning of biological tissue
Durable with prolonged sharpness, DURAEDGE® blades are finely sharpened, honed, and polished. They are then PTFE or Ceramic coated to reduce friction, section compression, and eliminate surface striations. Compatible with most blade holders. Made of high quality hardened stainless steel.
Certifications: CE Marked. Made in the USA.
Ordering information: Convenient 50 blade dispenser safely stores used blades.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together