Mini Magnifier, Heathrow Scientific®
Catalog # 82008-002
Supplier: Heathrow Scientific
Specifications
- Description:Mini Magnifier
- Cat. no.:82008-002
- W×D:185×100 mm
- Supplier No.:HS15092D
Specifications
About this item
The Mini Magnifier enlarges objects for easier viewing
The magnifier is equipped with a swiveling, molded lens that provides 1.75X magnification. The magnifier stand is 207 mm high.
Ordering information: Mini LED light box must be purchased separately.