VWR® Buffer Solutions | Avantor
VWR® Buffer Solutions
Supplier:   VWR
VWR® Buffer Solutions
Supplier:  VWR
Certificates
VWR® Buffer Solutions
VWR® Buffer Solutions
5 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These buffer solutions are for calibrating pH meters
They are traceable to NIST standards.