KIMBLE® KONTES® Universal Inlet with Hose Connection, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Designed to adapt a variety of tubes, condensers and filters in 14/20 and 24/25–24/40 glassware.
- Inert O-ring
- Vacuum tight seal
- Serrated side port
- Bottom inner joint
Diameter variations of 1 to 2mm can be accommodated, as the inert O-ring is compressed by the open-top polypropylene cap to effect a vacuum tight seal. The serrated side port can be used for a vacuum connection or for the introduction of gases. Bottom has an inner joint. Body is PTFE, O-rings are Viton, polypropylene compression cap is suitable for use to 150ºC, and side port is CTFE.
Ordering information: Includes Compression Cap.
Frequently Bought Together