Simple Machines, Block and Tackle Model | Avantor
Simple Machines, Block and Tackle Model
Simple Machines, Block and Tackle Model
Catalog # 470210-828
Supplier:   Wards
Fournisseur #:  WDMS21
Simple Machines, Block and Tackle Model
Catalog # 470210-828
Supplier:  Wards
Simple Machines, Block and Tackle Model
Simple Machines, Block and Tackle Model
Simple Machines, Block and Tackle Model
Price
87,50 $
Log in pour la disponibilité
Each
Product Details & Documents
This wooden machine has a sturdy wooden construction ideal for classroom use.
  • Demonstrate the use of pulleys
  • Includes manual
This apparatus can model how the use of several pulleys increases one's mechanical advantage. Students can use this both qualitatively and quantitatively and its design lends itself easily to inquiry based activities. Includes an instruction manual to facilitate student and teacher use of the apparatus.
Spécifications
  • Désignation:
    Simple Machines, Block and Tackle Model