ContactUsLinkComponent
Simple Machines, Fulcrum Balance Model
Catalog # 470210-820
Supplier:  Wards
undefined
Simple Machines, Fulcrum Balance Model
Catalog # 470210-820
Supplier:  Wards
Supplier Number:  WDMS17
Specifications
  • Description:
    Simple Machines, Fulcrum Balance Model
  • Cat. no.:
    470210-820
  • Supplier No.:
    WDMS17
Specifications
About this item
This wooden machine is elegantly designed and easy for students to use. Study how changing the distance from the fulcrum changes the mechanical advantage.
Students can intuitively figure out the balance point for a given set of masses. Includes an instruction manual to facilitate student and teacher use of the apparatus.