Simple Machines, Simple Wedge Model | Avantor
Simple Machines, Simple Wedge Model
Simple Machines, Simple Wedge Model
Catalog # 470210-814
Supplier:   Wards
Fournisseur #:  WDMS14
Simple Machines, Simple Wedge Model
Catalog # 470210-814
Supplier:  Wards
Simple Machines, Simple Wedge Model
Simple Machines, Simple Wedge Model
Simple Machines, Simple Wedge Model
Liquidation
$42.60
Vente
Valable jusqu'au 31/12/2026
Log in pour la disponibilité
Each
Product Details & Documents
This wooden machine is elegantly designed to demonstrate how a wedge can increase mechanical advantage.
  • Demonstrate mechanical advantages
  • Includes manual
Easy for students to use and measure. Includes an instruction manual to facilitate student and teacher use of the apparatus.
Spécifications
  • Désignation:
    Simple Machines, Simple Wedge Model