Simple Machines, Inclined Plane/Cart Model
Catalog # 470210-806
Supplier: Wards
Specifications
- Description:Simple Machines, Inclined Plane/Cart Model
- Cat. no.:470210-806
- Supplier No.:WDMS10
Specifications
About this item
This wooden machine is elegantly designed and easy for students to use.
- Cart with built-in mass
- Additional mass can be added
- Includes manual
It comes with a built-in mass, but the cart has a shallow indent for adding additional mass if needed. Students can study several different simple machine concepts with this one apparatus. Includes an instruction manual to facilitate student and teacher use of the apparatus.