Buffer No. 3, Hardy Diagnostics
Catalog # CA89498-952
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Pack type:Bottle
- Description:Ubuffer No. 3, USP<81>
- Size:500ml
- Storage temperature:2-8C
- Cat. no.:CA89498-952
- Supplier No.:U303
Specifications
About this item
Buffer No. 3, USP<81>, 500ml fill, 500ml polypropylene bottle, order by the package of 10, by Hardy Diagnostics
Buffers to be used in USP <81> antibiotic assay testing.
Buffer No
3, USP<81>, 500ml fill, 500ml polypropylene bottle, order by the package of 10, by Hardy Diagnostics
Certifications: USP<81>