Programmable DC Power Supply
À propos de cet article
A must for the lab that needs precise voltage or current.
- Compatible with iPad or iPhone
- Delivers up to 1560 Watts of Power
This series of programmable DC power supplies can be controlled through your iPad or iPhone using the exclusive B&K Precision PwrApp, which can be downloaded for free from the App Store on your Apple device. It delivers up to 1560 watts of clean power on a single output. The compact 2 inch high 19 inch wide supplies were designed to meet the needs of both system integrators and bench top users. Fast average command processing times of less than 50 ms, combined with the flexibility of industry-standard GPIB, LAN, USB & RS485 interfaces make these supplies ideal for ATE applications. Bench top users will benefit from the built-in voltage and current meters displaying setting and output values concurrently, an intuitive user interface with full keypad and rotary knob as well as cost effective USB & RS485 interfaces that come standard with all models. 1.44 KW.
2 Options Available Below
Product Details & Documents
A must for the lab that needs precise voltage or current.
- Compatible with iPad or iPhone
- Delivers up to 1560 Watts of Power
This series of programmable DC power supplies can be controlled through your iPad or iPhone using the exclusive B&K Precision PwrApp, which can be downloaded for free from the App Store on your Apple device. It delivers up to 1560 watts of clean power on a single output. The compact 2 inch high 19 inch wide supplies were designed to meet the needs of both system integrators and bench top users. Fast average command processing times of less than 50 ms, combined with the flexibility of industry-standard GPIB, LAN, USB & RS485 interfaces make these supplies ideal for ATE applications. Bench top users will benefit from the built-in voltage and current meters displaying setting and output values concurrently, an intuitive user interface with full keypad and rotary knob as well as cost effective USB & RS485 interfaces that come standard with all models. 1.44 KW.
Product Family Options
Informations sur le produit
- DésignationOutput VoltageInput VoltageCurrentAvailabilityPrice
- Programmable power supply 80V/18A0 - 80 V DC110 V / 220 V0 - 18 A DCLog in pour la disponibilitéEach$4,480.34Supplier Number: XLN8018-GL
- Programmable power supply 36V/40A0 - 36 V DC110 V / 220 V0 - 40 A DCLog in pour la disponibilitéEach$4,274.44Supplier Number: XLN3640-GL